Assay Details
| Gene Symbol: Slc38a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009950.1 | NP_001009950.1 | 6 - 7 | 854 | 70 | |
| GenBank mRNA | BC089013.1 | - | 6 - 7 | 854 | 30605563 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a8
Genomic Map