Assay Details
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009462.1 | NP_033488.1 | 6 - 7 | 1449 | 71 | |
GenBank mRNA | AK077453.1 | - | 6 - 7 | 1471 | 71 | |
AK129083.1 | - | 6 - 7 | 1457 | 71 | ||
AK137015.1 | - | 6 - 7 | 1471 | 71 | ||
AK157970.1 | - | 6 - 7 | 1473 | 71 | ||
AK165156.1 | - | 6 - 7 | 1469 | 71 | ||
AK166243.1 | - | 6 - 7 | 1466 | 71 | ||
AK172443.1 | - | 6 - 7 | 1471 | 71 | ||
BC007134.1 | - | 6 - 7 | 1451 | 3491297 | 71 | |
D84096.1 | - | 6 - 7 | 1449 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
