Assay Details
| Gene Symbol: Tmc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195088.1 | NP_001182017.1 | 3 - 4 | 585 | 69 | |
| NM_001195089.1 | NP_001182018.1 | 3 - 4 | 484 | 69 | ||
| NM_001195090.1 | NP_001182019.1 | 3 - 4 | 484 | 69 | ||
| NM_181856.2 | NP_862904.1 | 3 - 4 | 484 | 69 | ||
| GenBank mRNA | AK143862.1 | - | 3 - 4 | 587 | 69 | |
| AK156764.1 | - | 3 - 4 | 445 | 69 | ||
| AK171772.1 | - | 3 - 4 | 467 | 69 | ||
| AY236501.1 | - | 3 - 4 | 396 | 69 | ||
| AY263160.1 | - | 2 - 3 | 372 | 69 | ||
| BC138254.1 | - | 2 - 3 | 360 | 8861273 | 69 | |
| BC145210.1 | - | 2 - 3 | 360 | 9053761 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmc8
Genomic Map