Assay Details
| Gene Symbol: Tmc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195088.1 | NP_001182017.1 | 4 - 5 | 729 | 130 | |
| NM_001195089.1 | NP_001182018.1 | 4 - 5 | 628 | 130 | ||
| GenBank mRNA | AK143862.1 | - | 4 - 5 | 132 | ||
| AK171772.1 | - | 4 - 5 | 611 | 130 | ||
| AK220211.1 | - | 1 - 2 | 97 | 130 | ||
| BC145210.1 | - | 3 - 4 | 504 | 9053761 | 130 | |
More Information
Set Membership: |
|

Tmc8
Genomic Map