| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100037514.mm.1 |
NM_153145.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037514.mm.1 |
NM_153145.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037514.mm.1 |
TC1100003999.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100037518.mm.1 |
NM_153145.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037518.mm.1 |
NM_153145.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037518.mm.1 |
TC1100003999.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100037519.mm.1 |
NM_153145.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037519.mm.1 |
NM_153145.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037519.mm.1 |
TC1100003999.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100037516.mm.1 |
NM_153145.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037516.mm.1 |
NM_153145.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100037516.mm.1 |
TC1100003999.mm.1 |
MTA TC |