Assay Details
Gene Symbol: Abca8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153145.4 | NP_694785.3 | 22 - 23 | 3109 | 89 | |
GenBank mRNA | AB255881.1 | - | 21 - 22 | 3028 | 89 | |
AB255882.1 | - | 22 - 23 | 3116 | 89 | ||
AF498360.1 | - | 21 - 22 | 3037 | 89 | ||
AK031843.1 | - | 3 - 4 | 340 | 89 | ||
AK148846.1 | - | 2 - 3 | 199 | 89 | ||
AY732492.1 | - | 21 - 22 | 3037 | 89 | ||
BC026496.1 | - | 2 - 3 | 234 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
