Assay Details
Gene Symbol: Abca8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153145.4 | NP_694785.3 | 27 - 28 | 3691 | 126 | |
GenBank mRNA | AB255881.1 | - | 26 - 27 | 3610 | 126 | |
AB255882.1 | - | 27 - 28 | 3698 | 126 | ||
AF498360.1 | - | 26 - 27 | 3619 | 126 | ||
AK031843.1 | - | 8 - 9 | 922 | 126 | ||
AY732492.1 | - | 26 - 27 | 3619 | 126 | ||
BC026496.1 | - | 7 - 8 | 816 | 126 | ||
BC060032.1 | - | 9 - 10 | 1185 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
