Assay Details
Gene Symbol: Abca8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153145.4 | NP_694785.3 | 30 - 31 | 4024 | 125 | |
GenBank mRNA | AB255881.1 | - | 29 - 30 | 3943 | 125 | |
AB255882.1 | - | 30 - 31 | 4031 | 125 | ||
AF498360.1 | - | 29 - 30 | 3952 | 125 | ||
AK031843.1 | - | 11 - 12 | 1255 | 125 | ||
AY732492.1 | - | 29 - 30 | 3952 | 125 | ||
BC026496.1 | - | 10 - 11 | 1149 | 125 | ||
BC060032.1 | - | 12 - 13 | 1518 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
