Assay Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085498.2 | NP_001078967.1 | 4 - 5 | 415 | 97 | |
| GenBank mRNA | AK172123.1 | - | 4 - 5 | 382 | 97 | |
| AK219882.1 | - | 2 - 3 | 143 | 97 | ||
| BC147754.1 | - | 4 - 5 | 410 | 9056703 | 97 | |
| BC147755.1 | - | 4 - 5 | 410 | 9056704 | 97 | |
| BG078687.2 | - | 4 - 5 | 330 | 97 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chmp6
Genomic Map