Assay Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085498.2 | NP_001078967.1 | 7 - 8 | 614 | 80 | |
| GenBank mRNA | AK162346.1 | - | 3 - 4 | 191 | 80 | |
| AK184598.1 | - | 2 - 3 | 114 | 80 | ||
| BC147754.1 | - | 7 - 8 | 609 | 9056703 | 80 | |
| BC147755.1 | - | 7 - 8 | 609 | 9056704 | 80 | |
| BG078687.2 | - | 7 - 8 | 529 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098300 | BC147754.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098300 | BC147755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098300 | BG078687.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098300 | AK172123.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098300 | NM_001085498.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121474 | BC027085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121474 | AK172123.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121474 | AK179032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121474 | AK211523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121474 | NM_001085498.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73165 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098300 | BC147754.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098300 | BC147755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098300 | BG078687.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098300 | AK172123.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098300 | NM_001085498.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121474 | BC027085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121474 | AK172123.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121474 | AK179032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121474 | AK211523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121474 | NM_001085498.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02445442 | - | - |
| Whole Mouse Genome G4122A | A_52_P162538 | BC147754.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162538 | BC147755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162538 | BG078687.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162538 | AK172123.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162538 | BB870252.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28180985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28181008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09068264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09370339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09375302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09068262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09490266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09370341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09524527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09248766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09080838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09080840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09375293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09490268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09248760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09375294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09371869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09237814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28180966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28181050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10383286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383283 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383284 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433684_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10383282 | - | - |
| Mouse Genome 430 2.0 Array | 1433684_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433684_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208092_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208092_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100019335.mm.1 | NM_001085498.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019335.mm.1 | NM_001085498.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019335.mm.1 | TC1100002013.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019333.mm.1 | NM_001085498.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019333.mm.1 | NM_001085498.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019333.mm.1 | TC1100002013.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019334.mm.1 | NM_001085498.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019334.mm.1 | NM_001085498.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019334.mm.1 | TC1100002013.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433684_at | - | - |
| Mouse Expression Set 430 | 1433684_PM_at | - | - |
| Murine Genome U74v2 Set | 96097_PM_at | - | - |
| Murine Genome U74v2 Set | 96097_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433684_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433684_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433684_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433684_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251583 | - | - |

Chmp6
Genomic Map