Assay Details
| Gene Symbol: Hhip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020259.4 | NP_064655.4 | 2 - 3 | 962 | 122 | |
| GenBank mRNA | AK031539.1 | - | 2 - 3 | 961 | 122 | |
| AK031841.1 | - | 2 - 3 | 472 | 122 | ||
| BC053012.1 | - | 2 - 3 | 947 | 6402422 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hhip
Genomic Map