Assay Details
| Gene Symbol: Gaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159324.1 | NP_001152796.1 | 18 - 19 | 2815 | 63 | |
| NM_008064.3 | NP_032090.3 | 18 - 19 | 2905 | 63 | ||
| GenBank mRNA | AK052211.1 | - | 18 - 19 | 2886 | 63 | |
| AK088481.1 | - | 18 - 19 | 2881 | 63 | ||
| AK139333.1 | - | 18 - 19 | 2853 | 63 | ||
| AK141546.1 | - | 18 - 19 | 2888 | 63 | ||
| AK145631.1 | - | 17 - 18 | 2322 | 63 | ||
| AK146538.1 | - | 18 - 19 | 2809 | 63 | ||
| AK150970.1 | - | 18 - 19 | 2863 | 63 | ||
| AK206628.1 | - | 1 - 2 | 46 | 63 | ||
| AK209790.1 | - | 1 - 2 | 93 | 63 | ||
| BC010210.1 | - | 18 - 19 | 2869 | 3969127 | 63 | |
| U49351.1 | - | 18 - 19 | 2757 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gaa
Genomic Map