Assay Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029074.3 | NP_083350.2 | 2 - 3 | 178 | 117 | |
| GenBank mRNA | AK017198.1 | - | 2 - 3 | 139 | 117 | |
| AK146578.1 | - | 2 - 3 | 169 | 117 | ||
| BC094385.1 | - | 2 - 3 | 137 | 117 | ||
| BC118981.1 | - | 2 - 3 | 139 | 117 | ||
| BC148291.2 | - | 2 - 3 | 139 | 40103934 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnep1r1
Genomic Map