Assay Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029074.3 | NP_083350.2 | 4 - 5 | 366 | 78 | |
| GenBank mRNA | AK146578.1 | - | 4 - 5 | 357 | 78 | |
| AK217813.1 | - | 1 - 2 | 96 | 78 | ||
| BC018498.1 | - | 3 - 4 | 264 | 78 | ||
| BC148291.2 | - | 4 - 5 | 327 | 40103934 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnep1r1
Genomic Map