Assay Details
| Gene Symbol: Smarcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130187.1 | NP_001123659.1 | 2 - 3 | 666 | 106 | |
| NM_031878.2 | NP_114084.2 | 1 - 2 | 300 | 106 | ||
| GenBank mRNA | AK159259.1 | - | 2 - 3 | 503 | 106 | |
| AK166123.1 | - | 2 - 3 | 459 | 106 | ||
| BC005732.1 | - | 1 - 2 | 290 | 2647494 | 106 | |
| CO425905.1 | - | 1 - 2 | 467 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK166123.1 transcripts.
Set Membership: |
|

Smarcd2
Genomic Map