Assay Details
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027740.6 | NP_082016.1 | 10 - 11 | 1153 | 62 | |
GenBank mRNA | AK016991.1 | - | 9 - 10 | 1077 | 62 | |
AK030180.1 | - | 9 - 10 | 1047 | 62 | ||
AK045302.1 | - | 10 - 11 | 1132 | 62 | ||
AK050233.1 | - | 10 - 11 | 1222 | 62 | ||
AK090075.1 | - | 10 - 11 | 1156 | 62 | ||
BC050888.1 | - | 9 - 10 | 1151 | 62 | ||
BC099926.1 | - | 10 - 11 | 1239 | 62 | ||
BC144819.1 | - | 11 - 12 | 1264 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
