Assay Details
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027740.6 | NP_082016.1 | 6 - 7 | 709 | 93 | |
GenBank mRNA | AK016991.1 | - | 5 - 6 | 633 | 93 | |
AK030180.1 | - | 5 - 6 | 603 | 93 | ||
AK045302.1 | - | 6 - 7 | 688 | 93 | ||
AK050233.1 | - | 6 - 7 | 778 | 93 | ||
AK090075.1 | - | 6 - 7 | 712 | 93 | ||
BC050888.1 | - | 5 - 6 | 707 | 93 | ||
BC099926.1 | - | 6 - 7 | 796 | 93 | ||
BC144819.1 | - | 6 - 7 | 798 | 93 | ||
BC145801.1 | - | 6 - 7 | 798 | 40131104 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
