Assay Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177771.5 | NP_808439.2 | 1 - 2 | 230 | 66 | |
| GenBank mRNA | AK173029.1 | - | 1 - 2 | 230 | 66 | |
| BC025563.1 | - | 1 - 2 | 226 | 5322893 | 66 | |
| BC065167.1 | - | 1 - 2 | 126 | 66 | ||
| BY195998.1 | - | 1 - 2 | 151 | 66 | ||
| BY248410.1 | - | 1 - 2 | 150 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025563.1 transcripts.
Set Membership: |
|

Klhl18
Genomic Map