Assay Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177771.5 | NP_808439.2 | 4 - 5 | 698 | 61 | |
| GenBank mRNA | AK044774.1 | - | 4 - 5 | 809 | 61 | |
| AK173029.1 | - | 4 - 5 | 698 | 61 | ||
| AK198584.1 | - | 1 - 2 | 50 | 61 | ||
| BC025563.1 | - | 4 - 5 | 694 | 5322893 | 61 | |
| CB247282.1 | - | 1 - 2 | 46 | 61 | ||
| CK619719.1 | - | 2 - 3 | 304 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl18
Genomic Map