Assay Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177771.5 | NP_808439.2 | 6 - 7 | 1001 | 76 | |
| GenBank mRNA | AK044774.1 | - | 6 - 7 | 1112 | 76 | |
| AK173029.1 | - | 6 - 7 | 1001 | 76 | ||
| BC065167.1 | - | 5 - 6 | 697 | 76 | ||
| CB247282.1 | - | 3 - 4 | 349 | 76 | ||
| CK619719.1 | - | 4 - 5 | 607 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klhl18
Genomic Map