Assay Details
| Gene Symbol: Ankrd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081379.2 | NP_001074848.2 | 10 - 11 | 7966 | 102 | |
| NR_037865.1 | 11 - 12 | 8063 | 102 | |||
| GenBank mRNA | BC059876.1 | - | 2 - 3 | 1879 | 102 | |
| BC158043.1 | - | 9 - 10 | 7631 | 9088159 | 102 | |
| BC172141.1 | - | 10 - 11 | 7728 | 9054811 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd11
Genomic Map