| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800023938.mm.1 |
NM_001081379.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023938.mm.1 |
NM_001081379.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023938.mm.1 |
NR_037865.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023938.mm.1 |
NR_037865.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023938.mm.1 |
TC0800003084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023941.mm.1 |
NM_001081379.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023941.mm.1 |
NM_001081379.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023941.mm.1 |
NR_037865.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023941.mm.1 |
NR_037865.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023941.mm.1 |
TC0800003084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023944.mm.1 |
NM_001081379.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023944.mm.1 |
NM_001081379.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023944.mm.1 |
NR_037865.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023944.mm.1 |
NR_037865.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023944.mm.1 |
TC0800003084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023939.mm.1 |
NM_001081379.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023939.mm.1 |
NM_001081379.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023939.mm.1 |
NR_037865.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023939.mm.1 |
NR_037865.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023939.mm.1 |
TC0800003084.mm.1 |
MTA TC |