Assay Details
| Gene Symbol: Ankrd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081379.2 | NP_001074848.2 | 3 - 4 | 543 | 70 | |
| NR_037865.1 | 3 - 4 | 543 | 70 | |||
| GenBank mRNA | BC005576.1 | - | 1 - 2 | 140 | 70 | |
| BC068270.1 | - | 2 - 3 | 203 | 70 | ||
| BC158043.1 | - | 2 - 3 | 208 | 9088159 | 70 | |
| BC172141.1 | - | 2 - 3 | 208 | 9054811 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC172141.1 transcripts.
Set Membership: |
|

Ankrd11
Genomic Map