Assay Details
| Gene Symbol: Ankrd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081379.2 | NP_001074848.2 | 4 - 5 | 685 | 75 | |
| GenBank mRNA | BC005576.1 | - | 2 - 3 | 282 | 75 | |
| BC158043.1 | - | 3 - 4 | 350 | 9088159 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd11
Genomic Map