Assay Details
| Gene Symbol: Ankrd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081379.2 | NP_001074848.2 | 7 - 8 | 1196 | 59 | |
| NR_037865.1 | 8 - 9 | 1293 | 59 | |||
| GenBank mRNA | AK164512.1 | - | 4 - 5 | 3634 | 59 | |
| BC005576.1 | - | 5 - 6 | 793 | 59 | ||
| BC172141.1 | - | 7 - 8 | 958 | 9054811 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd11
Genomic Map