Assay Details
| Gene Symbol: Ccdc113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172914.2 | NP_766502.1 | 6 - 7 | 856 | 74 | |
| GenBank mRNA | AK077130.1 | - | 6 - 7 | 857 | 74 | |
| BC060957.1 | - | 6 - 7 | 854 | 6705542 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc113
Genomic Map