Assay Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 11 - 12 | 1589 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 11 - 12 | 1589 | 81 | |
Set Membership: |
|