Assay Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 14 - 15 | 2144 | 85 | |
| GenBank mRNA | AY352586.2 | - | 14 - 15 | 2141 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Drc7
Genomic Map