Assay Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 2 - 3 | 261 | 65 | |
| GenBank mRNA | AY352586.2 | - | 2 - 3 | 258 | 65 | |
| BC117982.1 | - | 1 - 2 | 235 | 65 | ||
| BC117983.1 | - | 1 - 2 | 235 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Drc7
Genomic Map