Assay Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 3 - 4 | 432 | 82 | |
| GenBank mRNA | AY352586.2 | - | 3 - 4 | 429 | 82 | |
| BC117982.1 | - | 2 - 3 | 406 | 82 | ||
| BC117983.1 | - | 2 - 3 | 406 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Drc7
Genomic Map