Assay Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 4 - 5 | 557 | 63 | |
| GenBank mRNA | AK076786.1 | - | 4 - 5 | 552 | 63 | |
| AY352586.2 | - | 4 - 5 | 554 | 63 | ||
| BC117982.1 | - | 3 - 4 | 531 | 63 | ||
| BC117983.1 | - | 3 - 4 | 531 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Drc7
Genomic Map