Assay Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 5 - 6 | 747 | 78 | |
| GenBank mRNA | AK076786.1 | - | 5 - 6 | 742 | 78 | |
| AY352586.2 | - | 5 - 6 | 744 | 78 | ||
| BC117982.1 | - | 4 - 5 | 721 | 78 | ||
| BC117983.1 | - | 4 - 5 | 721 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Drc7
Genomic Map