Assay Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042715.3 | NP_001036180.2 | 7 - 8 | 1131 | 58 | |
| GenBank mRNA | AY352586.2 | - | 7 - 8 | 1128 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Drc7
Genomic Map