Assay Details
| Gene Symbol: Gabarapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026693.5 | NP_080969.2 | 2 - 3 | 206 | 71 | |
| GenBank mRNA | AF190644.1 | - | 2 - 3 | 205 | 71 | |
| AK002596.1 | - | 2 - 3 | 186 | 71 | ||
| AK049819.1 | - | 188 | 71 | |||
| AK134873.1 | - | 207 | 71 | |||
| BC026798.1 | - | 2 - 3 | 166 | 4036425 | 71 | |
| BC081436.1 | - | 2 - 3 | 191 | 6814986 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171005 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171008 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171007 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141709 | AK134873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141709 | BC026798.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141709 | BC081436.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141709 | AK013563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2141709 | NM_026693.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561376 | AK051884.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147526 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166756 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126684 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141709 | AK134873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141709 | BC026798.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141709 | BC081436.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141709 | AK013563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2141709 | NM_026693.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517982 | AK134873.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517982 | AK184306.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517982 | BC026798.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517982 | AK013563.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517982 | AF190644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P2015 | AK049819.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26171006 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166756 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126684 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06643427 | - | - |
| Whole Mouse Genome G4122A | A_51_P517982 | AK134873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517982 | AK184306.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517982 | BC026798.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517982 | AK013563.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517982 | AF190644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P2015 | AK049819.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123629 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130243 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126209 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136178 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120103 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06643427 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126684 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166756 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171008 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26171005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26170980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26171009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07361016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06718853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06781884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07201074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07201078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06759201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26170991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07128111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07060228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26170997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26170993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07275535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07393636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06714282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26170989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06721414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26170999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06759202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26170988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07393633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07400958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06759199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06788050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26171005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07070489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07070498 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10575617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575618 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441376_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423187_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528546 | - | - |
| Mouse Genome 430 2.0 Array | 1423187_at | - | - |
| Mouse Genome 430 2.0 Array | 1423187_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441376_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441376_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93739_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93739_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800011046.mm.1 | NM_026693.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011046.mm.1 | NM_026693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011046.mm.1 | TC0800001341.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011044.mm.1 | NM_026693.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011044.mm.1 | NM_026693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011044.mm.1 | TC0800001341.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011050.mm.1 | NM_026693.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011050.mm.1 | NM_026693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011050.mm.1 | TC0800001341.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011052.mm.1 | NM_026693.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011052.mm.1 | NM_026693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011052.mm.1 | TC0800001341.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011051.mm.1 | NM_026693.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011051.mm.1 | NM_026693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011051.mm.1 | TC0800001341.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011048.mm.1 | NM_026693.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011048.mm.1 | NM_026693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011048.mm.1 | TC0800001341.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011053.mm.1 | NM_026693.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011053.mm.1 | NM_026693.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011053.mm.1 | TC0800001341.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423187_at | - | - |
| Mouse Expression Set 430 | 1423187_PM_at | - | - |
| Mouse Expression Set 430 | 1441376_PM_at | - | - |
| Mouse Expression Set 430 | 1441376_at | - | - |
| Murine Genome U74v2 Set | 96840_PM_at | - | - |
| Murine Genome U74v2 Set | 96840_at | - | - |
| Murine Genome U74v2 Set | 129072_at | - | - |
| Murine Genome U74v2 Set | 129072_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423187_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423187_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423187_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423187_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441376_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441376_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 381588 | - | - |
| ABI Mouse Genome Survey Microarray | 898845 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3360047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 610204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000610204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642197 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219431 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2642197 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219431 | - | - |
| mouse-6 v1.1 (Illumina) | scl093739.4_24-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0093739.1_283-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360047 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 610204 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0093739.1_283-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl093739.4_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610204 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2642197 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219431 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2642197 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219431 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0093739.1_283-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl093739.4_24-S | - | - |

Gabarapl2
Genomic Map