Assay Details
| Gene Symbol: Wfdc15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183271.2 | NP_899094.1 | 1 - 2 | 167 | 69 | |
| GenBank mRNA | AK006775.1 | - | 1 - 2 | 161 | 69 | |
| AK018854.1 | - | 1 - 2 | 161 | 69 | ||
| AK018888.1 | - | 1 - 2 | 168 | 69 | ||
| AK018909.1 | - | 1 - 2 | 139 | 69 | ||
| BC100423.1 | - | 1 - 2 | 146 | 30919644 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wfdc15a
Genomic Map