Assay Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028221.4 | NP_082497.2 | 2 - 3 | 328 | 87 | |
| GenBank mRNA | AK010053.1 | - | 2 - 3 | 293 | 87 | |
| AK033988.1 | - | 2 - 3 | 259 | 87 | ||
| AK050704.1 | - | 2 - 3 | 263 | 87 | ||
| BC016088.1 | - | 1 - 2 | 68 | 4503452 | 87 | |
| BC023158.1 | - | 2 - 3 | 167 | 4946201 | 87 | |
| BC025472.1 | - | 2 - 3 | 304 | 5353148 | 87 | |
| BC027060.1 | - | 2 - 3 | 290 | 5358022 | 87 | |
| BC027818.1 | - | 1 - 2 | 100 | 5320967 | 87 | |
| BC040818.1 | - | 281 | 4194531 | 87 | ||
| BY245102.1 | - | 3 - 4 | 387 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK033988.1,AK050704.1,BC016088.1,BC023158.1,BC025472.1,BC027060.1,BC027818.1,BC040818.1,NM_028221.4 transcripts.
This assay is in the 5'UTR region of the AK033988.1,AK050704.1,BC016088.1,BC023158.1,BC025472.1,BC027060.1,BC027818.1,BC040818.1,NM_028221.4 transcripts.
Set Membership: |
|

Fam192a
Genomic Map