Assay Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028221.4 | NP_082497.2 | 3 - 4 | 466 | 79 | |
| GenBank mRNA | AK010053.1 | - | 3 - 4 | 431 | 79 | |
| AK033988.1 | - | 3 - 4 | 397 | 79 | ||
| AK050704.1 | - | 3 - 4 | 401 | 79 | ||
| AK151603.1 | - | 3 - 4 | 420 | 79 | ||
| BC016088.1 | - | 2 - 3 | 206 | 4503452 | 79 | |
| BC023158.1 | - | 3 - 4 | 305 | 4946201 | 79 | |
| BC025472.1 | - | 3 - 4 | 442 | 5353148 | 79 | |
| BC027060.1 | - | 3 - 4 | 428 | 5358022 | 79 | |
| BC027818.1 | - | 2 - 3 | 234 | 5320967 | 79 | |
| BC040818.1 | - | 419 | 4194531 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC027818.1 transcripts.
Set Membership: |
|

Fam192a
Genomic Map