Assay Details
| Gene Symbol: Ucp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009463.3 | NP_033489.1 | 5 - 6 | 1049 | 73 | |
| GenBank mRNA | AK002759.1 | - | 5 - 6 | 1047 | 73 | |
| AK076454.1 | - | 5 - 6 | 1044 | 73 | ||
| AV248262.2 | - | 3 - 4 | 404 | 73 | ||
| BC012701.1 | - | 5 - 6 | 1046 | 4191276 | 73 | |
| U63419.1 | - | 5 - 6 | 1045 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426353 | U63419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P426353 | U63419.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57463 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155438 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426353 | U63419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P426353 | U63419.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426353 | U63419.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P426353 | U63419.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26014263 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01781812 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155438 | - | - |
| Whole Mouse Genome G4122A | A_51_P426353 | U63419.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P426353 | U63419.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129413 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155438 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07385070 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07385084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07385078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07385073 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07385067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26014307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07153155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07153154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06850232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07246704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07182595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06697230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07106410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07008197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07385069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06697231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07385075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07246711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07418517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07385074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07396028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07130291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26014324 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10573158 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573156 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418197_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573152 | - | - |
| Mouse Genome 430 2.0 Array | 1418197_at | - | - |
| Mouse Genome 430 2.0 Array | 1418197_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22227_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22227_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800006791.mm.1 | NM_009463.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006791.mm.1 | NM_009463.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006791.mm.1 | TC0800000919.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006792.mm.1 | NM_009463.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006792.mm.1 | NM_009463.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006792.mm.1 | TC0800000919.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 99507_at | - | - |
| Murine Genome U74v2 Set | 161422_PM_f_at | - | - |
| Murine Genome U74v2 Set | 99507_PM_at | - | - |
| Murine Genome U74v2 Set | 161422_f_at | - | - |
| Murine Genome U74v2 Set | 165911_r_at | - | - |
| Murine Genome U74v2 Set | 165911_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169349_at | - | - |
| Murine Genome U74v2 Set | 169349_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418197_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418197_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 932079 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6020368 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6020368 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020368 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2447464 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2447464 | - | - |
| mouse-6 v1.1 (Illumina) | scl33610.6.1_104-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020368 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33610.6.1_104-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020368 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2994373 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2994373 | - | - |

Ucp1
Genomic Map