Assay Details
| Gene Symbol: Tmem95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195710.1 | NP_001182639.1 | 1 - 2 | 175 | 91 | |
| GenBank mRNA | BF018485.1 | - | 1 - 2 | 122 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem95
Genomic Map