Assay Details
| Gene Symbol: Tmem74bos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042610.1 | - | 1 - 2 | 161 | 85 | |
| AK137371.1 | - | 1 - 2 | 162 | 85 | ||
| BC052486.1 | - | 1 - 2 | 381 | 85 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121857 | AK042610.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121857 | AK137371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P121857 | BC052486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121857 | AK042610.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121857 | AK137371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121857 | BC052486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183334 | AK042610.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183334 | AK137371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183334 | BC052486.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121857 | AK042610.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121857 | AK137371.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121857 | BC052486.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02135071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01938134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01792343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01593381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01173088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01920763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02135068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02135070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01938124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01461191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01173092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01683763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01173096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01461190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01151085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21745263 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 875569 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100580673 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2497028 | - | - |

Tmem74bos
Genomic Map