Assay Details
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008489.2 | NP_032515.2 | 12 - 13 | 1439 | 75 | |
| GenBank mRNA | BC004795.1 | - | 12 - 13 | 1327 | 3589072 | 75 |
| BQ176753.1 | - | 3 - 4 | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Lbp
Genomic Map