Assay Details
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008489.2 | NP_032515.2 | 1 - 2 | 302 | 91 | |
| GenBank mRNA | AK143562.1 | - | 1 - 2 | 303 | 91 | |
| AK158105.1 | - | 1 - 2 | 185 | 91 | ||
| BC004795.1 | - | 1 - 2 | 190 | 3589072 | 91 | |
| X99347.1 | - | 1 - 2 | 252 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lbp
Genomic Map