Assay Details
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008489.2 | NP_032515.2 | 3 - 4 | 548 | 74 | |
| GenBank mRNA | AK143562.1 | - | 3 - 4 | 549 | 74 | |
| AK158105.1 | - | 3 - 4 | 431 | 74 | ||
| BC004795.1 | - | 3 - 4 | 436 | 3589072 | 74 | |
| X99347.1 | - | 3 - 4 | 498 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lbp
Genomic Map