| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200021822.mm.1 |
NM_019547.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021822.mm.1 |
NM_019547.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021822.mm.1 |
TC0200002678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021829.mm.1 |
NM_019547.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021829.mm.1 |
NM_019547.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021829.mm.1 |
TC0200002678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021820.mm.1 |
NM_019547.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021820.mm.1 |
NM_019547.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021820.mm.1 |
TC0200002678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021821.mm.1 |
NM_019547.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021821.mm.1 |
NM_019547.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021821.mm.1 |
TC0200002678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021828.mm.1 |
NM_019547.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021828.mm.1 |
NM_019547.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021828.mm.1 |
TC0200002678.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021831.mm.1 |
NM_019547.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021831.mm.1 |
NM_019547.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021831.mm.1 |
TC0200002678.mm.1 |
MTA TC |