Assay Details
| Gene Symbol: Cct2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007636.2 | NP_031662.2 | 13 - 14 | 1443 | 61 | |
| GenBank mRNA | AB041570.1 | - | 13 - 14 | 1274 | 61 | |
| AK012295.1 | - | 13 - 14 | 1403 | 61 | ||
| AK075581.1 | - | 13 - 14 | 1416 | 61 | ||
| AK088145.1 | - | 13 - 14 | 1405 | 61 | ||
| AK145406.1 | - | 13 - 14 | 1444 | 61 | ||
| AK166891.1 | - | 13 - 14 | 1416 | 61 | ||
| BC007470.1 | - | 13 - 14 | 1407 | 3601569 | 61 | |
| BC026918.1 | - | 13 - 14 | 1409 | 4500896 | 61 | |
| Z31553.1 | - | 13 - 14 | 1394 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct2
Genomic Map