Assay Details
| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144534.1 | NP_653117.1 | 5 - 6 | 671 | 75 | |
| GenBank mRNA | AB261158.1 | - | 5 - 6 | 670 | 75 | |
| AK157969.1 | - | 5 - 6 | 731 | 75 | ||
| AK165818.1 | - | 6 - 7 | 731 | 75 | ||
| BC014728.1 | - | 5 - 6 | 684 | 4504294 | 75 | |
| BC024788.1 | - | 5 - 6 | 671 | 5360106 | 75 | |
| EF988666.1 | - | 5 - 6 | 705 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25962149 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25962150 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303725 | BC024788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303725 | NM_144534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303725 | BC024788.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P303725 | NM_144534.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430662 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943630 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938117 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27972 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176277 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143571 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303725 | BC024788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303725 | NM_144534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303725 | BC024788.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P303725 | NM_144534.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962162 | - | - |
| Whole Mouse Genome G4122A | A_52_P434729 | BC024788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434729 | AK165818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434729 | NM_144534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P434729 | AK003216.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303725 | BC024788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303725 | NM_144534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303725 | BC024788.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P303725 | NM_144534.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06931664 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25962150 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25962149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07246923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06810457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06872772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07296378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962176 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572818 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424177_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424178_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572815 | - | - |
| Mouse Genome 430 2.0 Array | 1424177_at | - | - |
| Mouse Genome 430 2.0 Array | 1424178_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424178_at | - | - |
| Mouse Genome 430 2.0 Array | 1424177_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74166_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74166_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800006122.mm.1 | NM_144534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006122.mm.1 | NM_144534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006122.mm.1 | TC0800000830.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006123.mm.1 | NM_144534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006123.mm.1 | NM_144534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006123.mm.1 | TC0800000830.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006121.mm.1 | NM_144534.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006121.mm.1 | NM_144534.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006121.mm.1 | TC0800000830.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424177_at | - | - |
| Mouse Expression Set 430 | 1424178_at | - | - |
| Mouse Expression Set 430 | 1424177_PM_at | - | - |
| Mouse Expression Set 430 | 1424178_PM_at | - | - |
| Murine Genome U74v2 Set | 167185_PM_i_at | - | - |
| Murine Genome U74v2 Set | 167185_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424177_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424178_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424178_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424177_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424178_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424177_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424178_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424177_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 624651 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 770114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 770114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770114 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245673 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245673 | - | - |
| mouse-6 v1.1 (Illumina) | scl074166.7_124-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl074166.7_124-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245673 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245673 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl074166.7_124-S | - | - |

Tmem38a
Genomic Map