Assay Details
| Gene Symbol: Mri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026423.4 | NP_080699.3 | 3 - 4 | 784 | 102 | |
| GenBank mRNA | AK007709.1 | - | 3 - 4 | 745 | 102 | |
| AK010538.1 | - | 3 - 4 | 756 | 102 | ||
| AK013882.1 | - | 3 - 4 | 753 | 102 | ||
| AK047287.1 | - | 3 - 4 | 102 | |||
| AK144807.1 | - | 3 - 4 | 727 | 102 | ||
| AK164894.1 | - | 3 - 4 | 753 | 102 | ||
| AK185486.1 | - | 1 - 2 | 93 | 102 | ||
| BC025049.1 | - | 3 - 4 | 751 | 4951045 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mri1
Genomic Map