Assay Details
| Gene Symbol: Banp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110100.2 | NP_001103570.1 | 6 - 7 | 884 | 56 | |
| NM_001285981.1 | NP_001272910.1 | 5 - 6 | 767 | 56 | ||
| NM_001285983.1 | NP_001272912.1 | 5 - 6 | 767 | 56 | ||
| NM_016812.4 | NP_058092.2 | 6 - 7 | 884 | 56 | ||
| GenBank mRNA | AF091234.1 | - | 5 - 6 | 678 | 56 | |
| AF235503.1 | - | 6 - 7 | 795 | 56 | ||
| AK154088.1 | - | 5 - 6 | 670 | 56 | ||
| AK170884.1 | - | 5 - 6 | 683 | 56 | ||
| BC021650.1 | - | 6 - 7 | 802 | 3660187 | 56 | |
| BC022168.1 | - | 6 - 7 | 802 | 4189797 | 56 | |
| BC062641.1 | - | 6 - 7 | 829 | 5718330 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Banp
Genomic Map