Assay Details
| Gene Symbol: Banp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285981.1 | NP_001272910.1 | 4 - 5 | 589 | 76 | |
| NM_001285983.1 | NP_001272912.1 | 4 - 5 | 589 | 76 | ||
| GenBank mRNA | AF091234.1 | - | 4 - 5 | 500 | 76 | |
| AK154088.1 | - | 4 - 5 | 492 | 76 | ||
| AK170884.1 | - | 4 - 5 | 504 | 76 | ||
| AK182116.1 | - | 1 - 2 | 165 | 76 | ||
| CX207431.1 | - | 4 - 5 | 589 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Banp
Genomic Map