Assay Details
| Gene Symbol: Dus2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301176.1 | NP_001288105.1 | 1 - 2 | 105 | 80 | |
| NM_025518.4 | NP_079794.1 | 1 - 2 | 105 | 80 | ||
| GenBank mRNA | AK009391.1 | - | 1 - 2 | 77 | 80 | |
| AK077451.1 | - | 1 - 2 | 77 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009391.1, NM_001301176.1, NM_025518.4 transcripts.
Set Membership: |
|

Dus2
Genomic Map